1-Benzylpyrrolidine-2,5-dione 95%
Lieferant: Apollo Scientific
OR949957-1G
OR949957-5G
APOSOR949957-1GEA
24.4
EUR
APOSOR949957-1G
APOSOR949957-5G
1-Benzylpyrrolidine-2,5-dione 95%
1-Benzylpyrrolidine-2,5-dione
Formel:
C₁₁H₁₁NO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00516834 CAS-Nummer: 2142-06-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...