3-Bromo-6-methoxypyridin-2-amine 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR949792-100MG
OR949792-250MG
APOSOR949792-100MGEA
45.1
EUR
APOSOR949792-100MG
APOSOR949792-250MG
3-Bromo-6-methoxypyridin-2-amine 97%
3-Bromo-6-methoxypyridin-2-amine
Formel:
C₆H₇BrN₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08062946 CAS-Nummer: 511541-63-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...