3-Propylisoxazol-5-amine 95%
Lieferant: Apollo Scientific
OR943659-1G
OR943659-250MG
APOSOR943659-1GEA
126
EUR
APOSOR943659-1G
APOSOR943659-250MG
3-Propylisoxazol-5-amine 95%
3-Propylisoxazol-5-amine
Formel:
C₆H₁₀N₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD05270911 CAS-Nummer: 747411-47-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...