N-Benzyl-2-methylpropan-1-amine 95%

Lieferant: Apollo Scientific

OR943596-1G OR943596-250MG OR943596-5G
APOSOR943596-1GEA 197 EUR
APOSOR943596-1G APOSOR943596-250MG APOSOR943596-5G
N-Benzyl-2-methylpropan-1-amine 95%
N-Benzyl-2-methylpropan-1-amine
Formel: C₁₁H₁₇N
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00972061
CAS-Nummer: 42882-36-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR