2,6-Dimethyl-3-nitroaniline 95%
Lieferant: Apollo Scientific
OR941380-1G
OR941380-5G
OR941380-25G
APOSOR941380-1GEA
45.1
EUR
APOSOR941380-1G
APOSOR941380-5G
APOSOR941380-25G
2,6-Dimethyl-3-nitroaniline 95%
2,6-Dimethyl-3-nitroaniline
Formel:
C₈H₁₀N₂O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00156348 CAS-Nummer: 67083-28-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...