1-(Trimethylacetyl)imidazole 95%

Lieferant: Apollo Scientific

OR940642-1G OR940642-5G
APOSOR940642-1GEA 105 EUR
APOSOR940642-1G APOSOR940642-5G
1-(Trimethylacetyl)imidazole 95%
1-(Trimethylacetyl)imidazole
Formel: C₈H₁₂N₂O
Molekulargewicht: 152.194 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07784427
CAS-Nummer: 4195-19-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR