2-(Methoxycarbonyl)isonicotinic acid 95%
Lieferant: Apollo Scientific
OR939996-1G
OR939996-250MG
APOSOR939996-1GEA
174
EUR
APOSOR939996-1G
APOSOR939996-250MG
2-(Methoxycarbonyl)isonicotinic acid 95%
2-(Methoxycarbonyl)isonicotinic acid
Formel:
C₈H₇NO₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00661398 CAS-Nummer: 24195-10-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...