3-(4-Hydroxyphenyl)-1-propanol 95%

Lieferant: Apollo Scientific

OR939367-1G OR939367-5G
APOSOR939367-1GEA 34.4 EUR
APOSOR939367-1G APOSOR939367-5G
3-(4-Hydroxyphenyl)-1-propanol 95%
3-(4-Hydroxyphenyl)-1-propanol
Formel: C₉H₁₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00002953
CAS-Nummer: 10210-17-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR