Ethyl 2-(cyclopropylsulphonyl)acetate 95%

Lieferant: Apollo Scientific

OR939223-1G OR939223-250MG
APOSOR939223-1GEA 632 EUR
APOSOR939223-1G APOSOR939223-250MG
Ethyl 2-(cyclopropylsulphonyl)acetate 95%
Ethyl 2-(cyclopropylsulphonyl)acetate
Formel: C₇H₁₂O₄S
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD20233368
CAS-Nummer: 1349719-17-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR