3,4-Dihydroxy-5-methoxybenzoic acid methyl ester 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-O-methylgallate, Methyl 3,4-dihydroxy-5-methoxybenzoate

OR939191-1G OR939191-5G
APOSOR939191-1GEA 172 EUR
APOSOR939191-1G APOSOR939191-5G
3,4-Dihydroxy-5-methoxybenzoic acid methyl ester 95%
3,4-Dihydroxy-5-methoxybenzoic acid methyl ester
Formel: C₉H₁₀O₅
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00183269
CAS-Nummer: 3934-86-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR