Ethyl 2-bromoacrylate 95%

Lieferant: Apollo Scientific

OR939045-1G OR939045-5G
APOSOR939045-1GEA 261 EUR
APOSOR939045-1G APOSOR939045-5G
Ethyl 2-bromoacrylate 95%
Ethyl 2-bromoacrylate
Formel: C₅H₇BrO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02258596
CAS-Nummer: 5459-35-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR