trans-4-Methoxy-β-nitrostyrol 96%
Lieferant: Apollo Scientific
OR936372-1G
OR936372-25G
OR936372-5G
APOSOR936372-1GEA
21.5
EUR
APOSOR936372-1G
APOSOR936372-25G
APOSOR936372-5G
trans-4-Methoxy-β-nitrostyrol 96%
trans-4-Methoxy-β-nitrostyrol
Formel:
C₉H₉NO₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00024826 CAS-Nummer: 5576-97-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...