2,5-Dimethyl-1H-indole-3-carbaldehyde 95%

Lieferant: Apollo Scientific

OR933305-1G OR933305-250MG
APOSOR933305-1GEA 712 EUR
APOSOR933305-1G APOSOR933305-250MG
2,5-Dimethyl-1H-indole-3-carbaldehyde 95%
2,5-Dimethyl-1H-indole-3-carbaldehyde
Formel: C₁₁H₁₁NO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06800613
CAS-Nummer: 61364-25-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR