2-Ethylnicotinic acid 95%

Lieferant: Apollo Scientific

OR932526-1G OR932526-5G
APOSOR932526-1GEA 98.8 EUR
APOSOR932526-1G APOSOR932526-5G
2-Ethylnicotinic acid 95%
2-Ethylnicotinic acid
Formel: C₈H₉NO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD18072523
CAS-Nummer: 3421-76-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR