Methyl benzooxazol-2-yl-acetate 95%

Lieferant: Apollo Scientific

OR931958-1G OR931958-250MG
APOSOR931958-1GEA 770 EUR
APOSOR931958-1G APOSOR931958-250MG
Methyl benzooxazol-2-yl-acetate 95%
Formel: C₁₀H₉NO₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11506177
CAS-Nummer: 75762-23-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR