Ethyl 4-aminopyridine-2-carboxylate 95%

Lieferant: Apollo Scientific

OR931425-1G OR931425-250MG
APOSOR931425-1GEA 609 EUR
APOSOR931425-1G APOSOR931425-250MG
Ethyl 4-aminopyridine-2-carboxylate 95%
Formel: C₈H₁₀N₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD10566236
CAS-Nummer: 773140-43-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR