3-(2-Propenyl)indole 95%
Lieferant: Apollo Scientific
Synonyme:
3-Allyl-1H-indole
OR930440-1G
OR930440-250MG
APOSOR930440-1GEA
435
EUR
APOSOR930440-1G
APOSOR930440-250MG
3-(2-Propenyl)indole 95%
3-(2-Propenyl)indole
Formel:
C₁₁H₁₁N Molekulargewicht: 157.212 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08361736 CAS-Nummer: 16886-09-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...