6-Chloro-N-methyl-3-pyridinamine 97%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-N-methylpyridin-3-amine

OR930363-1G OR930363-250MG
APOSOR930363-1GEA 112 EUR
APOSOR930363-1G APOSOR930363-250MG
6-Chloro-N-methyl-3-pyridinamine 97%
6-Chloro-N-methyl-3-pyridinamine
Formel: C₆H₇ClN₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD10697831
CAS-Nummer: 120739-84-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR