4-(Methylamino)cyclohexanone hydrochloride 96%
Lieferant: Apollo Scientific
OR927999-1G
OR927999-250MG
APOSOR927999-1GEA
429
EUR
APOSOR927999-1G
APOSOR927999-250MG
4-(Methylamino)cyclohexanone hydrochloride 96%
4-(Methylamino)cyclohexanone hydrochloride
Formel:
C₇H₁₃NO·ClH Molekulargewicht: 163.645 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD11520360 CAS-Nummer: 1260794-25-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...