4,4'-Diphenoxydiphenylsulphone 97%

Lieferant: Apollo Scientific

OR927070-1G OR927070-250MG
APOSOR927070-1GEA 300 EUR
APOSOR927070-1G APOSOR927070-250MG
4,4'-Diphenoxydiphenylsulphone 97%
4,4'-Diphenoxydiphenylsulphone
Formel: C₂₄H₁₈O₄S
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00221991
CAS-Nummer: 1623-91-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR