4-Cbz-amino-4-tetrahydropyrancarboxylic acid 97%

Lieferant: Apollo Scientific

OR926187-1G OR926187-500MG
APOSOR926187-1GEA 393 EUR
APOSOR926187-1G APOSOR926187-500MG
4-Cbz-amino-4-tetrahydropyrancarboxylic acid 97%
4-Cbz-amino-4-tetrahydropyrancarboxylic acid
Formel: C₁₄H₁₇NO₅
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD10698084
CAS-Nummer: 138402-13-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR