3,6-Dichloro-4,5-dimethylpyridazine 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR925462-100G
OR925462-25G
OR925462-5G
APOSOR925462-100GEA
186
EUR
APOSOR925462-100G
APOSOR925462-25G
APOSOR925462-5G
3,6-Dichloro-4,5-dimethylpyridazine 98%
3,6-Dichloro-4,5-dimethylpyridazine
Formel:
C₆H₆Cl₂N₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00067745 CAS-Nummer: 34584-69-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...