2-(Hydroxymethyl)benzonitrile 95%

Lieferant: Apollo Scientific

OR924435-1G OR924435-5G
APOSOR924435-1GEA 122 EUR
APOSOR924435-1G APOSOR924435-5G
2-(Hydroxymethyl)benzonitrile 95%
2-(Hydroxymethyl)benzonitrile
Formel: C₈H₇NO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11111887
CAS-Nummer: 89942-45-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR