2-Bromo-4-methyl-3-pentanone 95%

Lieferant: Apollo Scientific

OR923076-100MG OR923076-1G OR923076-250MG
APOSOR923076-100MGEA 154 EUR
APOSOR923076-100MG APOSOR923076-1G APOSOR923076-250MG
2-Bromo-4-methyl-3-pentanone 95%
2-Bromo-4-methyl-3-pentanone
Formel: C₆H₁₁BrO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD20621521
CAS-Nummer: 29583-93-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR