4-[(Methylamino)methyl]piperidine, N1-CBZ protected 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Benzyl 4-[(methylamino)methyl]piperidine-1-carboxylate
OR9218-50MG
OR9218-100MG
OR9218-2.5G
APOSOR9218-50MGEA
43.5
EUR
APOSOR9218-50MG
APOSOR9218-100MG
APOSOR9218-2.5G
4-[(Methylamino)methyl]piperidine, N1-CBZ protected 95%
4-[(Methylamino)methyl]piperidine, N1-CBZ protected
Formel:
C₁₅H₂₂N₂O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08271878 CAS-Nummer: 876316-35-7 UN-Nummer: 2922 ADR: 8+6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...