4-Amino-4-(hydroxymethyl)piperidine, N1-BOC protected 97+%

Lieferant: Apollo Scientific
Achtung

OR921438-1G OR921438-250MG
APOSOR921438-1GEA 580 EUR
APOSOR921438-1G APOSOR921438-250MG
4-Amino-4-(hydroxymethyl)piperidine, N1-BOC protected 97+%
4-Amino-4-(hydroxymethyl)piperidine, N1-BOC protected
Formel: C₁₁H₂₂N₂O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11845683
CAS-Nummer: 203186-96-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR