Methyl 3-hydroxycyclohexanecarboxylate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-Hydroxycyclohexanecarboxylate

OR921319-1G OR921319-5G
APOSOR921319-1GEA 60.9 EUR
APOSOR921319-1G APOSOR921319-5G
Methyl 3-hydroxycyclohexanecarboxylate 95%
Methyl 3-hydroxycyclohexanecarboxylate
Formel: C₈H₁₄O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11975826
CAS-Nummer: 37722-82-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR