(R)-Methyl 3-cyclopropyl-2-hydroxypropanoate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl (2R)-3-cyclopropyl-2-hydroxypropanoate

OR919615-250MG OR919615-1G
APOSOR919615-250MGEA 380 EUR
APOSOR919615-250MG APOSOR919615-1G
(R)-Methyl 3-cyclopropyl-2-hydroxypropanoate
(R)-Methyl 3-cyclopropyl-2-hydroxypropanoate
Formel: C₇H₁₂O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD22690391
CAS-Nummer: 1416444-91-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR