5-Bromo-1H-1,2,4-triazol-3-amine

Lieferant: Apollo Scientific

OR918830-1G OR918830-250MG
APOSOR918830-1GEA 85.7 EUR
APOSOR918830-1G APOSOR918830-250MG
5-Bromo-1H-1,2,4-triazol-3-amine
5-Bromo-1H-1,2,4-triazol-3-amine
Formel: C₂H₃BrN₄
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD18807020
CAS-Nummer: 389122-08-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR