1-(3-(tert-Butyl)phenyl)ethanone 96%

Lieferant: Apollo Scientific

OR918581-1G OR918581-5G
APOSOR918581-1GEA 31.9 EUR
APOSOR918581-1G APOSOR918581-5G
1-(3-(tert-Butyl)phenyl)ethanone 96%
1-(3-(tert-Butyl)phenyl)ethanone
Formel: C₁₂H₁₆O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11113485
CAS-Nummer: 6136-71-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR