1,1'-Carbonimidoylbis-1H-imidazole 95%
Lieferant: Apollo Scientific
Synonyme:
Di(1H-imidazol-1-yl)methanimine
OR917034-10G
OR917034-1G
OR917034-5G
APOSOR917034-10GEA
177
EUR
APOSOR917034-10G
APOSOR917034-1G
APOSOR917034-5G
1,1'-Carbonimidoylbis-1H-imidazole 95%
1,1'-Carbonimidoylbis-1H-imidazole
Formel:
C₇H₇N₅ Molekulargewicht: 161.164 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD10699388 CAS-Nummer: 104619-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...