1-Methylhistamine dihydrochloride 95%

Lieferant: Apollo Scientific
Gefahr

OR916820-1G OR916820-250MG OR916820-5G
APOSOR916820-1GEA 364 EUR
APOSOR916820-1G APOSOR916820-250MG APOSOR916820-5G
1-Methylhistamine dihydrochloride 95%
1-Methylhistamine dihydrochloride
Formel: C₆H₁₁N₃·2ClH
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00037000
CAS-Nummer: 6481-48-7
UN-Nummer: 2735
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR