Boc-d-homoserine

Lieferant: Apollo Scientific
Achtung

Synonyme: Boc-D-HoSer-OH

OR916500-1G OR916500-25G OR916500-5G
APOSOR916500-1GEA 74.1 EUR
APOSOR916500-1G APOSOR916500-25G APOSOR916500-5G
Boc-d-homoserine
Boc-d-homoserine
Formel: C₉H₁₇NO₅
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12545888
CAS-Nummer: 745011-75-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR