Boc-N-methyl-DL-alanine 97%
Lieferant: Apollo Scientific
Synonyme:
2-((tert-Butoxycarbonyl)(methyl)amino)propanoic acid
OR916415-25G
OR916415-5G
APOSOR916415-25GEA
132
EUR
APOSOR916415-25G
APOSOR916415-5G
Boc-N-methyl-DL-alanine 97%
Boc-N-methyl-DL-alanine
Formel:
C₉H₁₇NO₄ Molekulargewicht: 203.236 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00153297 CAS-Nummer: 13734-31-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...