(R)-3-(4-Chlorophenyl)-β-alaninol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3R)-3-Amino-3-(4-chlorophenyl)propan-1-ol

OR913669-1G OR913669-250MG
APOSOR913669-1GEA 877 EUR
APOSOR913669-1G APOSOR913669-250MG
(R)-3-(4-Chlorophenyl)-β-alaninol 97%
(R)-3-(4-Chlorophenyl)-β-alaninol
Formel: C₉H₁₂ClNO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07784070
CAS-Nummer: 1213362-28-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR