Ethyl 2-chloromethylbenzoate 95%

Lieferant: Apollo Scientific

OR911795-1G OR911795-250MG
APOSOR911795-1GEA 299 EUR
APOSOR911795-1G APOSOR911795-250MG
Ethyl 2-chloromethylbenzoate 95%
Ethyl 2-chloromethylbenzoate
Formel: C₁₀H₁₁ClO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00053935
CAS-Nummer: 1531-78-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR