5-Bromo-1,4-dimethylimidazole 98%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-1,4-dimethyl-1H-imidazole

OR910019-1G OR910019-250MG
APOSOR910019-1GEA 637 EUR
APOSOR910019-1G APOSOR910019-250MG
5-Bromo-1,4-dimethylimidazole 98%
5-Bromo-1,4-dimethylimidazole
Formel: C₅H₇BrN₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD23378484
CAS-Nummer: 861325-16-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR