6-Bromo-N-ethylpyridin-2-amine hydrochloride 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR909801-1G
OR909801-5G
APOSOR909801-1GEA
114
EUR
APOSOR909801-1G
APOSOR909801-5G
6-Bromo-N-ethylpyridin-2-amine hydrochloride 97%
6-Bromo-N-ethylpyridin-2-amine hydrochloride
Formel:
C₇H₉BrN₂·ClH Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD12198831 CAS-Nummer: 724770-74-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...