5,7-Dimethyl-1,3-benzothiazol-2-amine 95%
Lieferant: Apollo Scientific
OR909620-1G
OR909620-250MG
APOSOR909620-1GEA
435
EUR
APOSOR909620-1G
APOSOR909620-250MG
5,7-Dimethyl-1,3-benzothiazol-2-amine 95%
5,7-Dimethyl-1,3-benzothiazol-2-amine
Formel:
C₉H₁₀N₂S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04448825 CAS-Nummer: 73351-87-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...