5-(Methoxycarbonyl)furan-2-boronic acid 95%

Lieferant: Apollo Scientific

Synonyme: (5-(Methoxycarbonyl)furan-2-yl)boronic acid

OR908408-1G OR908408-250MG
APOSOR908408-1GEA 102 EUR
APOSOR908408-1G APOSOR908408-250MG
5-(Methoxycarbonyl)furan-2-boronic acid 95%
5-(Methoxycarbonyl)furan-2-boronic acid
Formel: C₆H₇BO₅
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD16660203
CAS-Nummer: 876189-20-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR