2-[(Dimethylamino)methyl]benzonitrile 97%

Lieferant: Apollo Scientific

OR908074-10G OR908074-1G OR908074-5G
APOSOR908074-10GEA 422 EUR
APOSOR908074-10G APOSOR908074-1G APOSOR908074-5G
2-[(Dimethylamino)methyl]benzonitrile 97%
2-[(Dimethylamino)methyl]benzonitrile
Formel: C₁₀H₁₂N₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07324818
CAS-Nummer: 53369-76-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR