4-Chloro-5-(methoxycarbonyl)thiophene-2-boronic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chloro-5-(methoxycarbonyl)thiophene-2-boronic acid
OR907381-250MG
OR907381-1G
APOSOR907381-250MGEA
475
EUR
APOSOR907381-250MG
APOSOR907381-1G
4-Chloro-5-(methoxycarbonyl)thiophene-2-boronic acid 97%
4-Chloro-5-(methoxycarbonyl)thiophene-2-boronic acid
Formel:
C₆H₆BClO₄S Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD18311836 CAS-Nummer: 2096331-56-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...