2-(4-Morpholino)-3-methylpyridine 97%

Lieferant: Apollo Scientific

OR902068-1G OR902068-5G
APOSOR902068-1GEA 145 EUR
APOSOR902068-1G APOSOR902068-5G
2-(4-Morpholino)-3-methylpyridine 97%
2-(4-Morpholino)-3-methylpyridine
Formel: C₁₀H₁₄N₂O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD10000144
CAS-Nummer: 327064-60-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR