1-BOC-4-Methylpyrazole-3-boronic acid, pinacol ester 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 4-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole-1-carboxylate
OR901385-250MG
OR901385-1G
APOSOR901385-250MGEA
607
EUR
APOSOR901385-250MG
APOSOR901385-1G
1-BOC-4-Methylpyrazole-3-boronic acid, pinacol ester 98%
1-BOC-4-Methylpyrazole-3-boronic acid, pinacol ester
Formel:
C₁₅H₂₅BN₂O₄ Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD21099686 CAS-Nummer: 1402174-62-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...