4-Hydroxy-N-propylbenzamide 98%

Lieferant: Apollo Scientific

OR900652-10G OR900652-1G OR900652-5G
APOSOR900652-10GEA 754 EUR
APOSOR900652-10G APOSOR900652-1G APOSOR900652-5G
4-Hydroxy-N-propylbenzamide 98%
4-Hydroxy-N-propylbenzamide
Formel: C₁₀H₁₃NO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06253037
CAS-Nummer: 27519-68-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR