4-Methyl-1-(methylamino)cyclohexane-1-carbonitrile hydrochloride 97%

Lieferant: Apollo Scientific

OR900488-1G OR900488-5G
APOSOR900488-1GEA 219 EUR
APOSOR900488-1G APOSOR900488-5G
4-Methyl-1-(methylamino)cyclohexane-1-carbonitrile hydrochloride 97%
4-Methyl-1-(methylamino)cyclohexane-1-carbonitrile hydrochloride
Formel: C₉H₁₆N₂·ClH
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD22689114
CAS-Nummer: 1420800-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR