tert-Butyl N-cyclohexylcarbamate 97%

Lieferant: Apollo Scientific

OR900319-1G OR900319-5G
APOSOR900319-1GEA 132 EUR
APOSOR900319-1G APOSOR900319-5G
tert-Butyl N-cyclohexylcarbamate 97%
tert-Butyl N-cyclohexylcarbamate
Formel: C₁₁H₂₁NO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07127722
CAS-Nummer: 3712-40-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR