4-(5-Bromothien-2-yl)pyrimidine-2-thiol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(5-Bromothiophen-2-yl)-2-sulphanylpyrimidine, 2-Bromo-5-(2-thiopyrimidin-4-yl)thiophene

OR6146-1G OR6146-5G
APOSOR6146-1GEA 152 EUR
APOSOR6146-1G APOSOR6146-5G
4-(5-Bromothien-2-yl)pyrimidine-2-thiol
4-(5-Bromothien-2-yl)pyrimidine-2-thiol
Formel: C₈H₅BrN₂S₂
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00084908
CAS-Nummer: 175202-82-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR