5-Bromo-4-methyl-1H-indazole, N1-BOC protected
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 5-bromo-4-methyl-1H-imidazole-1-carboxylate
OR60157-1G
OR60157-250MG
APOSOR60157-1GEA
140
EUR
APOSOR60157-1G
APOSOR60157-250MG
5-Bromo-4-methyl-1H-indazole, N1-BOC protected
5-Bromo-4-methyl-1H-indazole, N1-BOC protected
Formel:
C₁₃H₁₅BrN₂O₂ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD16628188 CAS-Nummer: 1257535-48-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...