2-Methylallylamine hydrochloride 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR59958-100G
OR59958-25G
OR59958-5G
APOSOR59958-100GEA
1220
EUR
APOSOR59958-100G
APOSOR59958-25G
APOSOR59958-5G
2-Methylallylamine hydrochloride 98%
2-Methylallylamine hydrochloride
Formel:
C₄H₉N·ClH Schmelzpunkt: 179…184 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00012876 CAS-Nummer: 28148-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...